Imprimer
Snap ring or crimp neck vials, E-Z Vial®, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
225179 225179-01 225179-03 225179-02
548-2106EA 241 EUR
548-2106 548-2108 548-2109 WHEA225179-02
Snap ring or crimp neck vials, E-Z Vial®, WHEATON®
Fioles
These snap ring vials made of clear or amber Type I borosilicate glass eliminate the need for crimping and decapping tools.
  • Vials accept snap cap or 11 mm aluminum seal
  • 40% larger opening improves sample accessibility while reducing auto sampler needle damage
Glass or PP vial inserts are available with or without writing patch.
Cdt: Vial inserts are packaged in shrink-wrapped trays of 100 for convenience and cleanliness.