Imprimer
Reagent bottles, safety coated, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
221014 221017
WHEA221014EA 133 EUR
WHEA221014 WHEA221017
Reagent bottles, safety coated, WHEATON®
Flacons Flacons en verre
Clear borosilicate glass with writing patch, Plastisol coated.
  • 33-430 screw cap attached
  • Polypropylene cap with PTFE liner
  • Autoclavable