Imprimer
Magnetic stirring bars, disposable, cylindrical, Spinbar®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F371120012 F371120001 F371120016 F371120002
BELAF371120012EA 357 EUR
BELAF371120012 BELAF371120001 BELAF371120016 BELAF371120002
Magnetic stirring bars, disposable, cylindrical, Spinbar®, SP Bel-Art
Agitateurs Barreaux magnétiques
Spinbar® magnetic stirring bars with Alnico V magnet encapsulated in FDA grade with PTFE. The multifaceted stirring bar surface adds turbulence relative to similar size smooth cylindrical stirring bars.
  • Disposable and eliminates potential for cross-contamination
  • Superior chemical compatibility
Ideal for single use applications such as mixing radioactive reagents where efficiency and cross-contamination are a concern.
Certifications: Manufactured in registered ISO 9001:2008 facilities, as verified by SGS certification.