Imprimer
Magnetic stirring bars, Pyrex® glass, Spinbar®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F371010001 F371010012 F371010112 F371017814
442-9412EA 135 EUR
442-9412 442-9410 442-9413 442-9411
Magnetic stirring bars, Pyrex® glass, Spinbar®, SP Bel-Art
Agitateurs Barreaux magnétiques
These Spinbar® Pyrex® glass encapsulated Alnico magnet stirring bars are useful for high temperature stirring up to 274 ˚C (525 ˚F).
  • Completely smooth surface offers zero absorption
  • Easy to clean
Certifications: Meets US Pharmacopoeia specifications for Type I Borosilicate glass.
Manufactured in registered ISO 9001:2008 facilities, as verified by SGS certification.