Imprimer
Magnetic stirring bar retrievers, flexible, PTFE, Spinbar®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H377740000 H377740101
BELAH377740000EA 0 EUR
BELAH377740000 BELAH377740101
Magnetic stirring bar retrievers, flexible, PTFE, Spinbar®, SP Bel-Art
Agitateurs Outils d'agitation
Flexible, PTFE rod with a powerful PTFE coated Alnico magnet for retrieving magnetic stirring bars from containers or sink drains and traps which are non-magnetic.
  • Flexible white rod is 330 mm long
Magnet length: 53 mm
Overall length: 383 mm