Imprimer
Bottle and vial racks, No-Wire™, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F185140025 F185140013 F185140016 F185140020 F185140030
212-8513EA 76.4 EUR
212-8513 212-8510 212-8511 212-8512 212-8514
Bottle and vial racks, No-Wire™, SP Bel-Art
Portoirs Portoirs pour flacons
Racks made of PP, white.
  • Five grid configuration
  • Holds small vials and bottles
  • Autoclavable
Small enough to hold in one hand and enough choices to accommodate most vials and small bottles.