Imprimer
Temperature probes, Lemo® compact, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
P1730-23 P1730-25 P1730-20 P1730-10
BOHLP1730-23EA 468 EUR
BOHLP1730-23 BOHLP1730-25 BOHLP1730-20 BOHLP1730-10
Temperature probes, Lemo® compact, BOLA
Sondes de température
Used for temperature measurement in aggressive liquids and ideal for built-in measurement cables.
  • Thermocouple in a PTFE-encapsulated stainless steel tube
  • Temperature probe with tip Ø 6 mm and collar ring Ø 12 mm
  • Connection by a coupling (type Lemo®, socket size 1, 4-pole) fixed directly at the end of the probe
  • Typical response times of T50 is 7 to 12 seconds and T90 is 14 to 16 seconds
Certifications: FDA conformed