Imprimer
VWR® Pop-Up™, Portoirs pour tubes à centrifuger, pliables
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
211-0368EA 56.5 EUR
211-0368 211-0367 525-0510 525-0511
VWR® Pop-Up™, Portoirs pour tubes à centrifuger, pliables
Portoirs Tube Racks
En PP
  • Simple easy action to open and close
  • Designed to help organise storage of 15 and 50 ml tubes
  • Stackable in open or closed position
  • Imprinted grid aids quick identification and retrieval
  • Collapse flat (height 21 mm) for compact storage
Close flat for storage and 'pop up' for quick use.
Note: Robust PP construction but not recommended for cleaning in an autoclave
Attention: Remarque: Construction robuste en PP, mais non recommandée pour l'autoclavage.