Imprimer
Pyruvaldéhyde, Liquid
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): Pyruvicaldehyde, Pyruvic aldehyde
0215555883 0215555825 0215555880
ICNA0215555883EA 270 EUR
ICNA0215555883 ICNA0215555825 ICNA0215555880
Pyruvaldéhyde, Liquid
Pyruvaldéhyde
Methylglyoxal is the aldehyde form of pyruvic acid
Methylglyoxal is used in formation of secondary organic aerosol. It it may be involved in the formation of advanced glycation endproducts (AGEs). Pyruvic aldehyde is used in the synthesis and cytotoxicity evaluation of N-terminal analogues of tubulysin-U an antimitotic compound. Also used in the synthesis of imidazoles as potent calcitonin gene-related CGRP antagonists.
Formule: C₃H₄O₂
Poids moléculaire: 72.06 g/mol
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00006960
Numéro CAS: 78-98-8
EINECS: 201-164-8