Imprimer
Wash bottles, wide neck, with coloured caps, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F116271000 F116250250 F116251000 F116260250 F116261000 F116270250 F116280250 F116281000 F116270500 F116201000
215-0311EA 0 EUR
215-0311 215-0305 215-0306 215-0308 215-0309 215-0310 215-0313 215-0314 BELAF116270500 BELAF116201000
Wash bottles, wide neck, with coloured caps, SP Bel-Art
Flacons Pissettes
LDPE, with colour coded PP spray caps.
  • Available with spray caps in five colours
  • Colour-coded closures for easy identification
  • Wide mouths for easy filling