Imprimer
Safety funnels with lid
Fournisseur: S.C.A.T.
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
107670 108670
SCTV107670EA 237 EUR
SCTV107670 SCTV108670
Safety funnels with lid
Entonnoirs Entonnoirs de sécurité
Safety funnels with hinged lid cap, sieve and safety lance.
  • Lid funnel with double thread R2" BSP/G2" (m) + 2" Tri-Sure (m) or S60/61
  • Made of PE-HD material
  • Stable stainless-steel strainer
  • Large funnel diameter: 250 mm
The funnels with hinged lid are suitable for universal use in the laboratory and are highly resistant to conventional chemicals due to the high-quality PE-Hd material.