Imprimer
Carboys, narrow neck, HDPE
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0402-0005 0402-0010 0402-0025 0402-0060
216-5712EA 59.5 EUR
216-5712 216-5714 216-5716 BURK0402-0060
Carboys, narrow neck, HDPE
Flacons Bonbonnes
Made of HDPE, translucent.
  • Blue printed graduation
  • Venting valve and other accessories must be ordered seperately
  • Good chemical and temperature resistance
Robust storage bottle with partcularly thick sides with sturdy carrying handle for safe trnasport and storage.