Imprimer
Funnels with long stem, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H920-06 H920-10 H920-04 H920-08 H920-02
BOHLH920-06EA 110 EUR
BOHLH920-06 221-1891 BOHLH920-04 BOHLH920-08 BOHLH920-02
Funnels with long stem, BOLA
Entonnoirs Entonnoirs pour la filtration Entonnoirs filtrants
Funnels, made of PTFE, white. Conical opening with long outlet.
  • Temperature resistance from −200 to +250 °C
  • Universal chemical resistance