Imprimer
Long ground sockets and cones, finished, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
286154602 286155804 286156603 286159206 286162602 286534404 286541605 286546209 286558408
SCOT286154602EA 332 EUR
SCOT286154602 SCOT286155804 SCOT286156603 SCOT286159206 SCOT286162602 SCOT286534404 SCOT286541605 SCOT286546209 SCOT286558408
Long ground sockets and cones, finished, DURAN®
Adaptateurs Raccords à rodage conique
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes