Imprimer
Kits d'amplification, illustra™ TempliPhi™
Marques : Cytiva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
25-6400-10 25-6400-50
25-6400-10EA 322 EUR
25-6400-10 25-6400-50
Kits d'amplification, illustra™ TempliPhi™
Réactifs pour acides nucléiques Isothermal Amplification Reagents
TempliPhi™ Kits efficiently prepare micrograms of circular DNA from picogram input material. The DNA templates are prepared by rolling circle amplification (RCA) using bacteriophage Phi29 DNA polymerase. TempliPhi™ uses an isothermal method for the exponential amplification of circular DNA. Phi29 DNA polymerase is active at 30 °C, enabling amplification to be performed at this temperature without the need for thermal cycling. The TempliPhi™ protocol requires less than 20 minutes of hands-on time to amplify 96 samples from bacterial colonies.
  • Prepare circular DNA templates for cycle sequencing, cloning, and transformation in 4 to 6 hours
  • Generate microgram quantities of template DNA from picogram amounts of starting material
  • Use amplified DNA directly for cycle sequencing without purification
  • Amplify DNA from bacterial or M13 liquid cultures, colonies, plaques, glycerol stocks, or purified circular (plasmid or M13) DNA
  • Simple protocol reduces time, labour, and consumables needed for template preparation, and workflow enables easy automation
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Application
Conditions de stockage 2 months or less at -20°C (for longer, store at -70°C). Thaw components on ice and maintain at 0–4°C during handling. DO NOT warm above 4°C prior to amplification.