Imprimer
Bottle top vacuum filtration systems, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N1650-24 N1650-16 N1650-08
BOHLN1650-24EA 6310 EUR
BOHLN1650-24 BOHLN1650-16 BOHLN1650-08
Bottle top vacuum filtration systems, BOLA
Filtres Unités de filtration sous vide
Vacuum filters with multi-stage hose connector with integrated lock screw for connecting vacuum tubing with Int.Ø 6 mm or 8 mm. The filters are produced without plasticisers and have an almost universal chemical resistance.
  • Collecting and filling vessel made of PFA
  • Filling vessel with lid for protection against contaminations
They do not release any trace elements into the filtrate. Due to the non-adhesive surface, they are easy to clean and can be reused.
Certifications: FDA conform.