Imprimer
Syringes for HPLC autosamplers, PerkinElmer instruments
Fournisseur: Trajan Scientific and Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
005990 008687 004995 006995 007995 008185
SGEA005990EA 113 EUR
SGEA005990 SGEA008687 SGEA004995 SGEA006995 SGEA007995 SGEA008185
Syringes for HPLC autosamplers, PerkinElmer instruments
Seringues Seringues de chromatographie
These syringes are made from borosilicate glass. They are used as replacement for C and CX syringes.
  • Accuracy and reproducibility of ±1%
  • Scale length: 60 mm
Certifications: Traceability to international standards.