Imprimer
Bottles, narrow neck, round, Boston type, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
W216845 W216846 W216842 W216849 W216836 W216837 W216838 W216839 W216840 W216841 W216848 W216850 W216851 W216852 W216853 W216843 W216800 W216835 W216844 W216847 W216803 W216808 W216807 W216801 W216802 W216804 W216805 W216806 W216809 W216810 W216811 W216812 W216813 W216814 W216815 W216816 W216817 W216832 W216867 W216834 W216869 W216870 W216833 W216823 W216822 W216821 W216831 W216866 W216868
WHEAW216845EA 29.1 EUR
WHEAW216845 WHEAW216846 WHEAW216842 WHEAW216849 WHEAW216836 WHEAW216837 WHEAW216838 WHEAW216839 WHEAW216840 WHEAW216841 WHEAW216848 WHEAW216850 WHEAW216851 WHEAW216852 WHEAW216853 WHEAW216843 WHEAW216800 WHEAW216835 WHEAW216844 WHEAW216847 WHEAW216803 WHEAW216808 WHEAW216807 WHEAW216801 WHEAW216802 WHEAW216804 WHEAW216805 WHEAW216806 WHEAW216809 WHEAW216810 WHEAW216811 WHEAW216812 WHEAW216813 WHEAW216814 WHEAW216815 WHEAW216816 WHEAW216817 WHEAW216832 WHEAW216867 WHEAW216834 WHEAW216869 WHEAW216870 WHEAW216833 WHEAW216823 WHEAW216822 WHEAW216821 WHEAW216831 WHEAW216866 WHEAW216868
Bottles, narrow neck, round, Boston type, WHEATON®
Flacons Flacons en verre
Soda lime glass, Class III, clear or amber.
  • Packaged in convenience packs with caps attached to maintain cleanliness
  • Polypropylene or phenolic caps with choice of liners
  • Hazardous communication label included with each case of bottles allows for content ID and safety notification
  • Tamper evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse
Ideal for solvent, chemical or sample storage. Amber coloured for light-sensitive materials.