Imprimer
Sample bottles, wide neck, with snap cap, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
225534 225535 225532 225536 225538 225543 225546
216-3071EA 225 EUR
216-3071 216-3072 216-3070 216-3073 216-3074 WHEA225543 WHEA225546
Sample bottles, wide neck, with snap cap, WHEATON®
Flacons Flacons en verre
Sample bottles made of soda-lime glass and pre-attached with LDPE snap caps.
Ideal for collecting and storing samples in the laboratory. These bottles were packaged in corrugated trays with partitions.
Certifications: Glass conforms to USP Type III requirements.