Imprimer
Bottles, narrow neck, round, LDPE, with screw caps, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
221140 221142 221143 221144
WHEA221140EA 152 EUR
WHEA221140 WHEA221142 WHEA221143 WHEA221144
Bottles, narrow neck, round, LDPE, with screw caps, WHEATON®
Flacons Flacons en plastique
LDPE, translucent.
  • Very flexible with high impact strength
  • Excellent for mild and strong buffers, good chemical resistance
  • Narrow mouth ideal for liquids
  • Pre-attached white polypropylene screw cap with foamed polyethylene liner