Imprimer
Fork wrenches, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
D647-08 D647-24
BOHLD647-08EA 125 EUR
BOHLD647-08 BOHLD647-24
Fork wrenches, BOLA
Wrenches
These glass fibre reinforced PA fork wrenches can be used for tightening or opening screw joints also at high working temperatures.
  • Good chemical resistance
  • Temperature resistant from −10 to +140 °C
  • Low weight reduces risk of injury or damage