Imprimer
General-purpose thermometers for flasks, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
B287-09 B287-06 B287-03
BOHLB287-09EA 293 EUR
BOHLB287-09 BOHLB287-06 BOHLB287-03
General-purpose thermometers for flasks, BOLA
Thermomètres Thermomètres analogiques
These thermometer made of PTFE jacketed glass. Suitable for 500 to 6000 ml reaction vessels.
  • Temperature resistance from 0 to +250 °C
  • Chemical resistance: +++ universal