Imprimer
Sample vials, with screw caps, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A195-04 A194-04
BOHLA195-04EA 88.8 EUR
BOHLA195-04 BOHLA194-04
Sample vials, with screw caps, BOLA
Fioles
These vials are suitable for centrifugal or evaporation purposes. They are made from PFA.
  • Temperature resistant from −200 to +250 °C
  • Chemical resistant