Imprimer
GC capillary column connectors, SilTite™ μ-unions
Fournisseur: Trajan Scientific and Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
073560 073561 073562 073563 073564 073565 073566 073567 073568 073569 073570 073571 073572 073573 073574
568-0287EA 187 EUR
568-0287 568-0288 568-0289 568-0290 568-0291 568-0292 568-0293 568-0294 568-0295 568-0296 568-0297 568-0298 568-0299 568-0300 568-0301
GC capillary column connectors, SilTite™ μ-unions
Fittings and Connectors
Ferrules for 0,1 to 0,25 mm ID columns have 0,4 mm holes. Ferrules for 0,32 mm ID columns have 0,5 mm holes. Ferrules for 0,53 mm ID columns have 0,8 mm holes.
  • Zero dead volume, ensuring optimised peak shapes
  • Leak-free FingerTite technology
  • Inert
  • Stable for both extreme temperature and pressure cycling
  • SilTite™ ferrules eliminate the need to re-tighten after installation
Informations de livraison: Each pack is delivered with two unions, five double-taper ferrules and the necessary tools.