Imprimer
Seringues haute pression
Fournisseur: Trajan Scientific and Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
008170 008171
SGEA008170EA 296 EUR
SGEA008170 SGEA008171
Seringues haute pression
Seringues Seringues de chromatographie
These high pressure syringes are designed for high pressure gas sampling.
  • Rated to 34 bar
  • Fitted with a gas tight on/off valve and high pressure 70 mm, 23 gauge needle
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable