Imprimer
Headspace/soil gas syringes
Fournisseur: Trajan Scientific and Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
008775 008975 031571
SGEA008775EA 269 EUR
SGEA008775 SGEA008975 SGEA031571
Headspace/soil gas syringes
Syringes Syringes for Chromatography
The gas tight syringes are fitted with a valve (SVLL) and a special Luer probe with a side hole for efficient sample intake.