Imprimer
Condensers, Liebig, vertical, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
B301-04 B301-02
BOHLB301-04EA 1060 EUR
BOHLB301-04 BOHLB301-02
Condensers, Liebig, vertical, BOLA
Condenseurs
PFA, PTFE, PP and glass.
  • Temperature resistance from −200 to +250 °C
  • Very good chemical resistance
  • Transparency: transparent
Thin-walled cooling tube made of translucent and gastight PFA, cooling jacket made of borosilicate glass with hose connectors made of PP and nuts for connection of cooling water, ground joint cone and socket made of PTFE. The distillate is only exposed to PFA/PTFE. Suitable for vertical assembly. Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.