Imprimer
Condensers, Liebig, vacuum, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
B295-02 B295-04
BOHLB295-02EA 803 EUR
BOHLB295-02 BOHLB295-04
Condensers, Liebig, vacuum, BOLA
Condenseurs
PFA, PTFE, PP and glass.
  • Temperature resistance from −200 to +250 °C
  • Very good chemical resistance
  • Transparency: transparent
One-piece cooling tube with ground joint socket and cone made of PTFE, cooling jacket made of borosilicate glass with hose connectors made of PP and nuts for connection of cooling water. The distillate is only exposed to PFA/PTFE. Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable