Imprimer
Glove liners, Perfect Poly® PU First White
Fournisseur: HONEYWELL SÉCURITÉ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2132255/10 2132255/11 2132255/6 2132255/7 2132255/8 2132255/9
DALL2132255/10EA 17.8 EUR
DALL2132255/10 DALL2132255/11 DALL2132255/6 DALL2132255/7 DALL2132255/8 DALL2132255/9
Glove liners, Perfect Poly® PU First White
Gants
100% Nylon knitted glove with white PU coating on the palm and fingers.
  • Ventilated back optimises the glove's breathability
  • PU coating ensures a good grip at the fingertips
  • White colour enhances the glove's hygiene and cleanliness
Ideal for fine handling or as an underglove.
Certifications: EN 388:2016 (3131X), Cat. II.