Imprimer
Boîtes de Petri, carrées, Sterilin®
Fournisseur: Sterilin
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
103-17 109-17
391-2017EA 829 EUR
391-2017 391-2018
Boîtes de Petri, carrées, Sterilin®
Boîtes de culture Boîtes de Petri
Les boîtes non compartimentées sont idéales pour l'analyse de sensibilité aux antibiotiques impliquant une très grande surface et une base très plate.
  • Ideal for antibiotic sensitivity testing when a large surface area and very flat base is required
  • Ideal for small volume liquid media work or for sample storage
  • Lid features selectable venting or non-venting positions where the lid turns through 90° to allow for venting or non-venting of the dish
Compartmentalised dish is ideal for small volume liquid media work or for sample storage. The 25 compartments each have a surface area of 1,8 cm² and a capacity of 5 ml. The lid features selectable venting or non venting positions.
Dimensions shown are for outer Ø base and outer height (mm).
Conduct various applications with Thermo Scientific™ Sterilin™ 100 mm square Petri dishes, available in both compartmentalised and non-compartmentalised formats. Aseptically manufactured under clean room conditions (class 7 ISO14644) to exclude microbiological contamination. Not available in America.