Imprimer
Chrome (III) oxyde ≥99.6% (base métaux), grumeau fondu 3-6mm
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): Chromic oxide
36258.14 36258.22
36258.14EA 67.5 EUR
36258.14 36258.22
Chrome (III) oxyde ≥99.6% (base métaux), grumeau fondu 3-6mm
Chrome (III) oxyde
For vacuum deposition
It can be used in selective oxidation. It is mainly used for smelting metal chromium and chromium carbide. It is used as coloring agent for color painting on enamelware and porcelain ware, imitation leather or building material. It is also used for making sun-proof coating, abrasive, green polishing compound and special printing ink for printing bank note. It is used as catalyst for organic synthetic chromium.
For vacuum deposition
Formule: Cr₂O₃
Poids moléculaire: 151.99 g/mol
Point d'ébullition: 3000 °C
Point de fusion: 2435
Densité: 5.22
Température de stockage: Température ambiante
Numéro MDL: MFCD00010949
Numéro CAS: 1308-38-9
EINECS: 215-160-9
Indice Merck: 14,2234