Chrome (III) oxyde 98+%, en poudre -325 Mesh
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Attention
|
It is employed in metallurgical processing and has applications in chrome plating, anodizing aluminum etc... It is also used as a pigment.
Purity: 98+%
CAS No.: 1308-38-9
Molecular Formula: Cr2O3
Formula Weight: 151.99
Physical Form: -325 Mesh Powder
Density: 5.22
Refractive Index: 2.551
Appearance: Pale to dark green
Odour: Odourless
Melting Point: 2435°
Boiling Point: 3000°
Solubility: Practically insoluble in water, alcohol, acetone. Slightly soluble in acids, alkalies
Merck Reference: 14,2234
MDL No.: MFCD00010949
Formule:
Cr₂O₃ Point d'ébullition: 3000 °C Point de fusion: 2435 °C Densité: 5,22 Température de stockage: Température ambiante |
Numéro MDL:
MFCD00010949 Numéro CAS: 1308-38-9 EINECS: 215-160-9 Indice Merck: 14,2234 |