Imprimer
Argent nitrate ≥99.9% (base métaux) ACS
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
11414.06 11414.14 11414.22 11414.36 11414.62 11414.A1
11414.06EA 55.8 EUR
11414.06 11414.14 11414.22 11414.36 11414.62 11414.A1
Argent nitrate ≥99.9% (base métaux) ACS
Argent nitrate
Purity: 99.9+% CAS No.: 7761-88-8 Molecular Formula: AgNO3 Formula Weight: 169.87 Physical Form: Crystalline Density: 4.352 Appearance: White Odour: Odourless Melting Point: 212° Solubility: Freely soluble in water, alcohol, ammonia water. Slightly soluble in ether Merck Reference: 14,8518 UN No.: 1479 MDL No.: MFCD00003414
Silver plating, photography, manufacturing of other silver compounds, mirrors, as an analytical lab reagent, in Colouring porcelain
Formule: AgNO₃
Point d'ébullition: 444 °C (1013 hPa)
Point de fusion: 212 °C
Densité: 4,352
Température de stockage: Température ambiante
Numéro MDL: MFCD00003414
Numéro CAS: 7761-88-8
EINECS: 231-853-9
UN: 1479
ADR: 5.1,I
REACH: 01-2119513705-43
Indice Merck: 14,8518
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM