Antimoine (III) chlorure ≥99.0% ACS
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Danger
|
Purity: 99.0% min
CAS No.: 10025-91-9
Molecular Formula: SbCl3
Formula Weight: 228.11
Physical Form: Crystalline
Density: 3.14
Melting Point: 73.4°
Boiling Point: 223.5°
Solubility: Soluble in water with hydrolysis to SbOCl. Soluble in HCl solution, alcohol, benzene, carbon disulfide, dioxane, chloroform, ether, acetone, carbon tetrachloride. Insoluble in organic bases
Merck Reference: 14,707
UN No.: 1733
MDL No.: MFCD00011212
Stripping tin coating from steel surfaces, in organic synthesis, as catalyst, in making other antimony salts, in chemical microscopy for drug identification
Formule:
SbCl₃ Point d'ébullition: 223,5 °C Densité: 3,14 Température de stockage: Température ambiante |
Numéro MDL:
MFCD00011212 Numéro CAS: 10025-91-9 EINECS: 233-047-2 UN: 1733 ADR: 8,II Indice Merck: 14,707 |