Imprimer
Etain (II) chlorure, anhydre ≥99%
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
Synonyme(s): Stannous chloride, Tin dichloride
41960.30 41960.A1 41960.A7
41960.30EA 127 EUR
41960.30 41960.A1 41960.A7
Etain (II) chlorure, anhydre ≥99%
Etain (II) chlorure
Purity: 99% min CAS No.: 7772-99-8 Molecular Formula: SnCl2 Formula Weight: 189.62 Physical Form: Crystalline Density: 3.95 Melting Point: 247° Boiling Point: 652° Solubility: Soluble in water, alkalies, alcohol, MEK, methyl acetate, ether, and acetone Merck Reference: 14,8783 UN No.: 3260 MDL No.: MFCD00011241
As a reducing agent, in plating reactions, as a galvanizing reagent, an antisludging agent, a stabilizer, and as a catalyst
Formule: SnCl₂
Poids moléculaire: 189.62 g/mol
Point d'ébullition: 652 °C
Point de fusion: 247 °C
Densité: 3.95
Température de stockage: Température ambiante
Numéro MDL: MFCD00011241
Numéro CAS: 7772-99-8
EINECS: 231-868-0
UN: 3260
ADR: 8 III
Indice Merck: 14,8783