Imprimer
Dishes
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
625 627
KART625EA 56.4 EUR
KART625 KART627
Dishes
Plateaux/Récipients/Pots Capsules d'évaporation
Made of PTFE, white.
  • Very good chemical resistance
  • Very good temperature resistance
  • Good mechanical resistance
Dishes are suitable for foodstuff. It can withstand temperatures continuously up to 250 °C and for short intervals up to 300 °C.