Imprimer
Drip pans
Fournisseur: USBECK Laborgeräte
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4781 4784
USBE4781EA 155 EUR
USBE4781 USBE4784
Drip pans
Plateaux/Récipients/Pots Plateaux de déversements
Made of stainless steel.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean
With removable gridiron.