Sodium hydroxyde, anhydre ≥97.0% ACS faible teneur en chlorure
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Danger
|
Synonyme(s):
Hydroxyde de sodium
Purity: 97.0% min
CAS No.: 1310-73-2
Molecular Formula: NaOH
Formula Weight: 40.00
Physical Form: Pellets
Density: 2.13
pH: 14
Melting Point: 318°
Boiling Point: 1390°
Solubility: Soluble in water, alcohol and glycerol
Merck Reference: 14,8627
UN No.: 1823
MDL No.: MFCD00003548
In chemical manufacture, rayon and cellophane, as a neutralizing agent in petroleum refining, in pulp, paper detergents and textiles, and as a lab reagent. NaOH solutions precipitate most metals as hydroxides from aqueous solutions of the metal salts
Formule:
NaOH Poids moléculaire: 40 g/mol Point d'ébullition: 1390 °C Point de fusion: 318 °C Densité: 2.13 Température de stockage: Température ambiante |
Numéro MDL:
MFCD00003548 Numéro CAS: 1310-73-2 EINECS: 215-185-5 UN: 1823 ADR: 8 II REACH: 01-2119457892-27 Indice Merck: 14,8627 |