Imprimer
Tubing connectors, straight, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
D575-08 D575-06 D575-04 D575-02
BOHLD575-08EA 21.3 EUR
BOHLD575-08 BOHLD575-06 BOHLD575-04 BOHLD575-02
Tubing connectors, straight, BOLA
Raccords et connecteurs
Straight fitting, made of PTFE, with two connectors for elastic tubing (e.g. Viton®, Tygon®, silicone).
  • Universal chemical resistance
  • Maximum temperature up to 250 °C
  • Suitable for vacuum