Imprimer
Tubing connectors, elbow-shaped, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
D574-06 D574-02 D574-08 D574-04
BOHLD574-06EA 36.3 EUR
BOHLD574-06 BOHLD574-02 BOHLD574-08 BOHLD574-04
Tubing connectors, elbow-shaped, BOLA
Raccords et connecteurs
Elbow-shaped fitting, made of PTFE, with two connectors for elastic tubing.
  • Universal chemical resistance
  • Temperature resistance -200 up to 250 °C
  • Vacuum: suitable