Imprimer
Screw caps for bottles with GL threads, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H986-04 H986-02 H986-03 H986-05 H986-01
BOHLH986-04EA 68.6 EUR
BOHLH986-04 BOHLH986-02 BOHLH986-03 BOHLH986-05 BOHLH986-01
Screw caps for bottles with GL threads, BOLA
Capuchons Bouchons pour flacons
Screw cap with handy knurl, made of PTFE.
  • Very good chemical resistance
  • Temperature resistant from −200 to +250 °C
Suitable for bottles and vessels with GL threads.