Imprimer
Multiple distributors with GL thread necks for canisters, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
D766-22 D762-08 D762-20 D762-32 D762-14 D762-38 D762-44 D766-16
BOHLD766-22EA 145 EUR
BOHLD766-22 BOHLD762-08 BOHLD762-20 BOHLD762-32 BOHLD762-14 BOHLD762-38 BOHLD762-44 BOHLD766-16
Multiple distributors with GL thread necks for canisters, BOLA
Capuchons Bouchons pour flacons
Screw cap made from PE and body made from PTFE.
  • Temperature resistance: –50 to +80 °C (* temperature resistance from –20 to +110 °C)
  • Very good chemical resistance