Imprimer

Eau, HyClone™ HyPure, qualité Biologie Moléculaire Molecular biology grade

Marques : HyClone products (Cytiva)
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Synonyme(s): Hydrogen oxide

SH30538.01 SH30538.02 SH30538.03 SH30538.FS SH30538.LS
SH30538.01EA 18.6 EUR
SH30538.01 HYCLSH30538.02 HYCLSH30538.03 HYCLSH30538.FS HYCLSH30538.LS
Eau, HyClone™ HyPure, qualité Biologie Moléculaire Molecular biology grade
Eau

Nuclease-free HyClone Molecular Biology-Grade Water, containing no DEPC or other carcinogenic chemicals is used for critical applications involving nucleotides such as RNA and DNA.


  • Significantly low levels of endotoxins
  • Certified to have no detectable activity in DNase, RNase, and protease assays
  • Maximum allowable limit of endotoxin is 0.05 EU/mL


HyClone HyPure Molecular Biology Grade Water purification process includes continuous deionization, reverse osmosis, ultraviolet light treatment, distillation, and 0.1 µm filtration.


Certifications: Manufactured using ISO 9001 and ISO 13485 certified processes according to 21 CFR 820 medical device current good manufacturing practices (cGMP).filtration

Formule: H₂O
Poids moléculaire: 18.02 g/mol
Point d'ébullition: 100
Point de fusion: 0
Densité: 1 g/cm³ (4 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00011332
Numéro CAS: 7732-18-5
EINECS: 231-791-2
Indice Merck: 13,10098

Commander maintenant

Résultats des tests de spécification

Endotoxin Concentration ≤ 0,05 EU/mL
Sterilisation 0,1 µm sterile filtered
pH 5,0 to 7,0
Storage 2° to 30° C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR