Potassium permanganate ≥99.0% ACS
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Danger
|
Synonyme(s):
Permanganate de potassium
Purity: 99.0% min
CAS No.: 7722-64-7
Molecular Formula: KMnO4
Formula Weight: 158.03
Physical Form: Crystalline
Density: 2.7
Appearance: Dark purple
Melting Point: 240° dec.
Solubility: Partially soluble in water. Decomposes in organic solvents and acids
Merck Reference: 14,7655
UN No.: 1490
MDL No.: MFCD00011364
In photography, tanning leather, purifying water, manufacturing mineral water, as an analytical lab reagent
Formule:
KMnO₄ Poids moléculaire: 158.03 g/mol Point de fusion: 240 °C (déc.) Densité: 2.7 Température de stockage: Température ambiante |
Numéro MDL:
MFCD00011364 Numéro CAS: 7722-64-7 EINECS: 231-760-3 UN: 1490 ADR: 5.1 II Indice Merck: 14,7655 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...