Imprimer
Stopcocks with hose connectors
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
E650-53 E650-56 E650-59 E650-03 E650-12 E650-09 E650-50 E650-06
BOHLE650-53EA 216 EUR
BOHLE650-53 BOHLE650-56 BOHLE650-59 BOHLE650-03 BOHLE650-12 BOHLE650-09 BOHLE650-50 BOHLE650-06
Stopcocks with hose connectors
Stopcocks
Made of PTFE.
  • Temperature resistance from −200 to 250 °C
  • Universal chemical resistance
  • Pressure upto 2 bar
Stopcocks with hose connectors for distributing liquids or gases, quick and easy disconnection of flow.
Two-way stopcock with straight bore and two hose connectors or three-way stopcock with T-shaped bore and three hose connectors for connecting elastic tubing. Conical stopcock plug, tightness is increased by turning the nut on the lower side.