Imprimer
Overhead stirrer tools, double impulse stirrer shafts, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C391-28 C391-18
BOHLC391-28EA 1470 EUR
BOHLC391-28 BOHLC391-18
Overhead stirrer tools, double impulse stirrer shafts, BOLA
Agitateurs Outils d'agitation
Stirrer shafts made of PTFE-jacketed stainless steel and the two paddles arranged crosswise at 90° completely made of PTFE.
  • Suitable for stirring in viscous liquids
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C
Upper paddle is fixed by means of clamp screws made of PEEK compound.