Imprimer
Watch dishes, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A200-02 A200-04 A200-03 A200-01
BOHLA200-02EA 15.7 EUR
BOHLA200-02 BOHLA200-04 BOHLA200-03 BOHLA200-01
Watch dishes, BOLA
Watch Glasses
Dish, made of PTFE, white.
  • Convex shape
  • Universal chemical resistance
  • Temperature resistance: from −200 to +250 °C
For blends or for covering vessels.