Imprimer

Potassium chlorate ≥99.0% ACS

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Danger

Synonyme(s): Chlorate de potassium

36494.36 36494.A3
36494.36EA 102 EUR
36494.36 36494.A3
Potassium chlorate ≥99.0% ACS
Potassium chlorate

Purity: 99.0% min CAS No.: 3811-04-9 Molecular Formula: KClO3 Formula Weight: 122.55 Physical Form: Crystalline Density: 2.32 Appearance: White Odour: Odourless Melting Point: 368° Boiling Point: 400° dec. Solubility: Soluble in water, glycerol. Insoluble in alcohol Merck Reference: 14,7620 UN No.: 1485 MDL No.: MFCD00011361


As an oxidizing agent, in textile printing and dyeing, as a disinfectant, in manufacturing of aniline black and other dyes

Formule: KClO₃
Poids moléculaire: 122.55 g/mol
Point d'ébullition: 400 °C (déc.)
Point de fusion: 368 °C
Densité: 2.32
Température de stockage: Température ambiante
Numéro MDL: MFCD00011361
Numéro CAS: 3811-04-9
EINECS: 223-289-7
UN: 1485
ADR: 5.1 II
Indice Merck: 14,7620

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR