Imprimer
Bottles, wide neck, PTFE, conical neck, with screw cap, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A111-32 A111-40 A111-24 A111-16
BOHLA111-32EA 129 EUR
BOHLA111-32 BOHLA111-40 BOHLA111-24 BOHLA111-16
Bottles, wide neck, PTFE, conical neck, with screw cap, BOLA
Flacons Flacons en plastique
PTFE, white.
  • Temperature resistance: from –200 to +250 °C
  • Universal chemical resistance
  • Pressure: no pressure
Thick-walled, smooth interior surface, ergonomic grips on bottle and screw cap.