Imprimer
Lithium chlorure ≥99% ACS
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
36217.36 36217.A3
36217.36EA 322 EUR
36217.36 36217.A3
Lithium chlorure ≥99% ACS
Lithium chlorure
Purity: 99% min CAS No.: 7447-41-8 Molecular Formula: LiCl Formula Weight: 42.39 Physical Form: Granular Density: 2.068 Melting Point: 605° Boiling Point: 1325-1360° Solubility: Very soluble in water, alcohol, ether, pyridine, and nitrobenzene Merck Reference: 14,5528 MDL No.: MFCD00011078
In air-conditioning, welding and soldering flux, dry batteries, heat-exchange media, salt baths, and desiccants
Formule: LiCl
Poids moléculaire: 42.39 g/mol
Point d'ébullition: 1325…1360 °C
Point de fusion: 605 °C
Densité: 2.068
Température de stockage: Température ambiante
Numéro MDL: MFCD00011078
Numéro CAS: 7447-41-8
EINECS: 231-212-3
Indice Merck: 14,5528